2-Chlor-5-iodpyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25271.03
B25271.06
B25271.14
B25271.03EA
33.4
EUR
B25271.03
B25271.06
B25271.14
2-Chlor-5-iodpyridin ≥98%
2-Chlor-5-iodpyridin
Purity: 98%
CAS No.: 69045-79-0
Molecular Formula: C5H3ClIN
Formula Weight: 239.44
Melting Point: 95-98°
Beilstein Reference: 108889
Formel:
C₅H₃ClIN Molekulargewicht: 239.44 g/mol Schmelzpunkt: 95…98 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01863635 CAS-Nummer: 69045-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...