2-Chlor-4-iodpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
378420050
378420250
ACRO378420050EA
46.8
EUR
ACRO378420050
ACRO378420250
2-Chlor-4-iodpyridin 98%
2-Chlor-4-iodpyridin
Formel:
C₅H₃ClIN Schmelzpunkt: 41…45 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01861983 CAS-Nummer: 153034-86-7 UN-Nummer: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to brown-yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 41 to 45 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...