2-Amino-6-brompyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine
M-Clarity™ quality level = MQ200
521744-25G
521744-5G
SIAL521744-25GEA
90.6
EUR
SIAL521744-25G
SIAL521744-5G
2-Amino-6-brompyridin, Sigma-Aldrich®
2-Amino-6-brompyridin
Formel:
C₅H₅BrN₂ |
MDL Number:
MFCD00137843 CAS-Nummer: 19798-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...