2-Brom-5-iodpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L20016.03
L20016.06
L20016.03EA
29.2
EUR
L20016.03
L20016.06
2-Brom-5-iodpyridin ≥97%
2-Brom-5-iodpyridin
Purity: 97%
CAS No.: 73290-22-9
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Melting Point: 123-124°
Beilstein Reference: 109100
Formel:
C₅H₃BrIN Molekulargewicht: 283.89 g/mol Schmelzpunkt: 123…124 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03095201 CAS-Nummer: 73290-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...