Bernsteinsäureanhydrid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydrofuran-2,5-dione
Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.
Formel:
C₄H₄O₃ Siedepunkt: 261 °C Schmelzpunkt: 118…121 °C Dichte: 1.572 Flammpunkt: 157 °C (314 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005525 CAS-Nummer: 108-30-5 EINECS: 203-570-0 UN-Nummer: 3261 ADR: 8,III Merck Index: 14,8870 |
Spezifikation Testergebnisse
Appearance (Color) | White |
Form | Powder or crystals or crystalline powder or flakes and/or chips |
Assay (Aqueous acidbase Titration) | ≥98.5 to ≤101.5% |
Identification (FTIR) | Conforms |
Melting Point (clear melt) | 116 - 121 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...