Pentadecan-15-olid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone
Purity: 98%
CAS No.: 106-02-5
Molecular Formula: C15H28O2
Formula Weight: 240.39
Flash Point:
Formel:
C₁₅H₂₈O₂ Siedepunkt: 137 °C (2 mmHg) Schmelzpunkt: 33…37 °C Dichte: 0.918 Flammpunkt: >110 °C (230 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00039667 CAS-Nummer: 106-02-5 EINECS: 203-354-6 Merck Index: 14,3910 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...