Eisen(II)sulfid ≥99,98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Ferrous sulphide

22388.06 22388.14
22388.06EA 142 EUR
22388.06 22388.14
Eisen(II)sulfid ≥99,98% (Metall-Basis)
Eisen(II)sulfid

Purity: 99.98% CAS No.: 1317-37-9 Molecular Formula: FeS Formula Weight: 87.91 Physical Form: Powder Density: 4.74 Appearance: Brown to black Odour: Odourless Melting Point: 1195° Merck Reference: 14,4058 MDL No.: MFCD00011013

Formel: FeS
Schmelzpunkt: 1195 °C
Dichte: 4.74
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011013
CAS-Nummer: 1317-37-9
EINECS: 215-268-6
UN-Nummer: 3077
ADR: 9,III
Merck Index: 14,4058

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR