Eisen(III)oxid ≥98% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12375.A1
12375.A7
12375.C4
12375.A1EA
66.3
EUR
12375.A1
12375.A7
12375.C4
Eisen(III)oxid ≥98% (Metall-Basis)
Eisen(III)oxid
Purity: 98%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: -325 Mesh Powder
Density: 5.24
Appearance: Red-brown
Odour: Odourless
Melting Point: 1565°
Merck Reference: 14,4028
MDL No.: MFCD00011008
Formel:
Fe₂O₃ Schmelzpunkt: 1565 °C Dichte: 5.24 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011008 CAS-Nummer: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...