Nickel(II)iodid, wasserfrei ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Nickel (II) iodide, Nickel iodide (NiI2)

22893.06 22893.14
22893.06EA 63.1 EUR
22893.06 22893.14
Nickel(II)iodid, wasserfrei ≥99,5% (Metall-Basis)
Nickel(II)iodid

Purity: 99.5% CAS No.: 13462-90-3 Molecular Formula: NiI2 Formula Weight: 312.51 Physical Form: -8 Mesh Density: 5.834 Melting Point: 797° UN No.: 3077 MDL No.: MFCD00016264

Formel: I₂Ni
Schmelzpunkt: 797 °C
Dichte: 5.834
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016264
CAS-Nummer: 13462-90-3
EINECS: 236-666-6
UN-Nummer: 3077
ADR: 9,III
Merck Index: 12,06594

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR