Rhodium(III)chlorid, wasserfrei ≥99,9% (Metall-Basis) Rh ≥ 48.7%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Rhodium(III) chloride, Rhodium trichloride
11815.01
11815.03
11815.06
11815.01EA
363
EUR
11815.01
11815.03
11815.06
Rhodium(III)chlorid, wasserfrei ≥99,9% (Metall-Basis) Rh ≥ 48.7%
Rhodium(III)chlorid
Purity: 99.9%
CAS No.: 10049-07-7
Molecular Formula: Cl3Rh
Formula Weight: 209.26
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Red-brown
Odour: Odourless
Merck Reference: 14,8188
MDL No.: MFCD00011204
Formel:
Cl₃Rh Dichte: 5.38 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011204 CAS-Nummer: 10049-07-7 EINECS: 233-165-4 Merck Index: 14,8188 |