Palladium(II)bromid 39,8-40,1% Pd

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Palladium dibromide, Palladium bromide

195190020 195190100
ACRO195190020EA 270 EUR
ACRO195190020 ACRO195190100
Palladium(II)bromid 39,8-40,1% Pd
Palladium(II)bromid
Formel: Br₂Pd
Dichte: 5,173 g/cm³ (16 °C)
MDL Number: MFCD00011170
CAS-Nummer: 13444-94-5
EINECS: 236-588-2

Bestellen

Spezifikation Testergebnisse

Appearance Dark red-brown to black Powder or crystals
Assay 39.8 to 40.1 % (Pd)
Calcium (Ca) ≤20 ppm
Copper (Cu) ≤10 ppm
Gold (Au) ≤20 ppm
Iridium (Ir) ≤100 ppm
Iron (Fe) ≤20 ppm
Magnesium (Mg) ≤10 ppm
Platinum (Pt) ≤100 ppm
Rhodium (Rh) ≤100 ppm
Ruthenium (Ru) ≤100 ppm
Silicon (Si) ≤50 ppm
Silver (Ag) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR