Nickel(II)bromid, wasserfrei 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Nickel dibromide
53115.18
53115.30
53115.18EA
128
EUR
53115.18
53115.30
Nickel(II)bromid, wasserfrei 99%
Nickel(II)bromid
Purity: 99%
CAS No.: 13462-88-9
Molecular Formula: Br2Ni
Formula Weight: 218.53
Physical Form: Crystalline
Density: 5.098
Melting Point: 963°
Merck Reference: 14,6502
UN No.: 3077
MDL No.: MFCD00011141
Formel:
Br₂Ni Schmelzpunkt: 963 °C Dichte: 5.098 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011141 CAS-Nummer: 13462-88-9 EINECS: 236-665-0 UN-Nummer: 3077 ADR: 9,III Merck Index: 14,6502 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...