Mangan(III)oxid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Manganese oxide (Mn2O3), Dimanganese trioxide

M-Clarity™ quality level = MQ200

377457-1KG 377457-250G
SIAL377457-1KGEA 465 EUR
SIAL377457-1KG SIAL377457-250G
Mangan(III)oxid, Sigma-Aldrich®
Mangan(III)oxid
Formel: Mn₂O₃
Schmelzpunkt: 1347 °C
Dichte: 4.81 g/cm³ (20 °C)
MDL Number: MFCD00016217
CAS-Nummer: 1317-34-6
EINECS: 215-264-4
Merck Index: 13,05761

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR