Mangan(II)iodid, wasserfrei 98%

Lieferant: Thermo Fisher Scientific
Achtung

32808.09 32808.18
32808.09EA 105 EUR
32808.09 32808.18
Mangan(II)iodid, wasserfrei 98%
Mangan(II)iodid

Purity: 98% CAS No.: 7790-33-2 Molecular Formula: I2Mn Formula Weight: 308.75 Physical Form: Powder Density: 5.0 Appearance: White to pink to red Odour: Odourless Melting Point: 638° Solubility: Soluble in water with gradual decomposition. Soluble in alcohol Merck Reference: 14,5732 MDL No.: MFCD00016225

Formel: I₂Mn
Schmelzpunkt: 638 °C
Dichte: 5,0
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016225
CAS-Nummer: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR