Vanadium(V)oxid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Vanadium pentoxide, di-Vanadium pentaoxide
M-Clarity™ quality level = MQ200
223794-100G
223794-500G
SIAL223794-100GEA
38.6
EUR
SIAL223794-100G
SIAL223794-500G
Vanadium(V)oxid, Sigma-Aldrich®
Vanadium(V)oxid
Formel:
O₅V₂ Siedepunkt: 1750 °C (1013 hPa) Schmelzpunkt: 690 °C Dichte: 3.35 g/cm³ (20 °C) |
MDL Number:
MFCD00011457 CAS-Nummer: 1314-62-1 EINECS: 215-239-8 UN-Nummer: 2862 ADR: 6.1,III Merck Index: 13,09987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...