Niob(V)oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diniobium pentaoxide, Niobium pentoxide

M-Clarity™ quality level = MQ100

208515-100G 208515-500G
SIAL208515-100GEA 196 EUR
SIAL208515-100G SIAL208515-500G
Niob(V)oxid, Sigma-Aldrich®
Niob(V)oxid
Formel: Nb₂O₅
Schmelzpunkt: 1496 °C
Dichte: 4.6 g/cm³ (20 °C)
MDL Number: MFCD00011128
CAS-Nummer: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR