Niob(V)oxid ≥99,95% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Diniobium pentaoxide, Niobium pentoxide
36478.14
36478.22
36478.14EA
120
EUR
36478.14
36478.22
Niob(V)oxid ≥99,95% (Metall-Basis)
Niob(V)oxid
Purity: 99.95%
CAS No.: 1313-96-8
Molecular Formula: Nb2O5
Formula Weight: 265.81
Physical Form: 3-12mm Sintered Lump
Density: 4.6
Melting Point: 1520°
Merck Reference: 14,6559
MDL No.: MFCD00011128
For vacuum deposition
Formel:
Nb₂O₅ Schmelzpunkt: 1520 °C Dichte: 4.6 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011128 CAS-Nummer: 1313-96-8 EINECS: 215-213-6 Merck Index: 14,6559 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...