Vanadium(III)fluorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Vanadium trifluoride

81115.04 81115.09 81115.18
81115.04EA 155 EUR
81115.04 81115.09 81115.18
Vanadium(III)fluorid ≥98%
Vanadium(III)fluorid

Purity: 98% CAS No.: 10049-12-4 Molecular Formula: VF3 Formula Weight: 107.94 Physical Form: Powder Density: 3.363 Melting Point: ≈1406° Solubility: Practically insoluble in water, acetone, and glacial acetic acid Merck Reference: 14,9922 UN No.: 2923 MDL No.: MFCD00042553

Formel: F₃V
Molekulargewicht: 107.94 g/mol
Schmelzpunkt: ∼1406 °C
Dichte: 3.363
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00042553
CAS-Nummer: 10049-12-4
EINECS: 233-169-6
UN-Nummer: 2923
ADR: 8 II
Merck Index: 12,10058

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR