Titan(II)hydrid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Titanium hydride, Titanium dihydride
M-Clarity™ quality level = MQ200
1.12384.0100
1.12384.1000
1.12384.0100EA
154
EUR
1.12384.0100
1.12384.1000
Titan(II)hydrid, Sigma-Aldrich®
Titan(II)hydrid
Formel:
H₂Ti Dichte: 3.91 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151301 CAS-Nummer: 7704-98-5 EINECS: 231-726-8 UN-Nummer: 1871 ADR: 4.1, Merck Index: 14,09473 |
Spezifikation Testergebnisse
Melting Point |
|
Molar Mass | 49.88 g/mol |
Bulk Density | 1400 kg/m3 |
Flash Point |
|
Density | 3.76 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 400 °C |
Hazard Class | 4.1 |
Hazard Class Description | Flammable Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...