Titan(II)hydrid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Titanium hydride, Titanium dihydride

89183.06 89183.18 89183.30
89183.06EA 38 EUR
89183.06 89183.18 89183.30
Titan(II)hydrid ≥99% (Metall-Basis)
Titan(II)hydrid

Purity: 99% CAS No.: 7704-98-5 Molecular Formula: H2Ti Formula Weight: 49.92 Physical Form: 1-3 Micron Powder Density: 3.9 Appearance: Dark grey Odour: Odourless Melting Point: 450° dec. Merck Reference: 14,9473 UN No.: 1871 MDL No.: MFCD00151301


For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H2

Formel: H₂Ti
Schmelzpunkt: 450 °C (dec.)
Dichte: 3.9
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151301
CAS-Nummer: 7704-98-5
EINECS: 231-726-8
UN-Nummer: 1871
ADR: 4.1,II
Merck Index: 14,9473

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR