Diyttriumtrisulfid ≥99,9%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Yttrium (III) sulphide, Yttrium sulphide
39565.03
39565.06
39565.14
39565.03EA
69.4
EUR
39565.03
39565.06
39565.14
Diyttriumtrisulfid ≥99,9%
Diyttriumtrisulfid
Purity: 99.9%
CAS No.: 12039-19-9
Molecular Formula: Y2S3
Formula Weight: 273.93
Physical Form: Powder
Odour: Like rotten eggs
UN No.: 3134
MDL No.: MFCD00058806
Formel:
S₃Y₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058806 CAS-Nummer: 12039-19-9 EINECS: 234-887-2 UN-Nummer: 3134 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...