Yttrium(III)fluorid, wasserfrei ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Yttrium trifluoride, Yttrium(III) fluoride
41424.09
41424.18
41424.30
41424.09EA
88.3
EUR
41424.09
41424.18
41424.30
Yttrium(III)fluorid, wasserfrei ≥99,9% (Metall-Basis)
Yttrium(III)fluorid
Purity: 99.9%
CAS No.: 13709-49-4
Molecular Formula: YF3
Formula Weight: 145.90
Physical Form: 3-12mm Pieces
Sensitivity: Hygroscopic
UN No.: 3288
MDL No.: MFCD00049714
For vacuum deposition
Formel:
F₃Y Molekulargewicht: 145.9 g/mol Siedepunkt: 2227 Schmelzpunkt: 1387 Dichte: 4.01 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00049714 CAS-Nummer: 13709-49-4 EINECS: 237-257-5 UN-Nummer: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...