Cadmium(II)iodid ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cadmium iodide
43442.22
43442.14
43442.22EA
518
EUR
43442.22
43442.14
Cadmium(II)iodid ≥99,999% (Metall-Basis), Puratronic®
Cadmium(II)iodid
Purity: 99.999%
CAS No.: 7790-80-9
Molecular Formula: CdI2
Formula Weight: 366.21
Physical Form: Powder
Density: 5.67
Appearance: Pale brown
Odour: Odourless
Melting Point: 387-389°
Boiling Point: 787°
Merck Reference: 14,1621
UN No.: 2570
MDL No.: MFCD00010920
Formel:
CdI₂ Siedepunkt: 787 °C Schmelzpunkt: 387…389 °C Dichte: 5.67 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010920 CAS-Nummer: 7790-80-9 EINECS: 232-223-6 UN-Nummer: 2570 ADR: 6.1,III Merck Index: 14,1621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...