Zinkchlorid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
Unter Argon in Ampullen verpackt
35726.06
35726.14
35726.22
35726.06EA
98.4
EUR
35726.06
35726.14
35726.22
Zinkchlorid, ultra trocken ≥99,999% (Metall-Basis)
Zinkchlorid
Purity: 99.999%
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.29
Physical Form: Powder, Ampouled under argon
Density: 2.91
Melting Point: 290°
Boiling Point: 732°
Merck Reference: 14,10132
UN No.: 2331
MDL No.: MFCD00011295
Formel:
ZnCl₂ Molekulargewicht: 136.3 g/mol Siedepunkt: 732 °C Schmelzpunkt: 290 °C Dichte: 2.91 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011295 CAS-Nummer: 7646-85-7 EINECS: 231-592-0 UN-Nummer: 2331 ADR: 8 III Merck Index: 14,10132 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...