Kupfer(II)oxid ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.0% min
CAS No.: 1317-38-0
Molecular Formula: CuO
Formula Weight: 79.55
Physical Form: Powder
Density: 6.3
Refractive Index: 2.63
Appearance: Black
Odour: Odourless
Melting Point: 1326°
Merck Reference: 14,2646
UN No.: 3077
MDL No.: MFCD00010979
Formel:
CuO Siedepunkt: 2000 °C (1013 hPa) Schmelzpunkt: 1326 °C Dichte: 6,3 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010979 CAS-Nummer: 1317-38-0 EINECS: 215-269-1 UN-Nummer: 3077 ADR: 9,III Merck Index: 14,2646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...