Silber(II)fluorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11610.04
11610.09
11610.18
11610.04EA
63.7
EUR
11610.04
11610.09
11610.18
Silber(II)fluorid ≥98%
Silber(II)fluorid
Purity: 98+%
CAS No.: 7783-95-1
Molecular Formula: AgF2
Formula Weight: 145.87
Physical Form: Crystalline
Density: 4.6
Appearance: Brown
Melting Point: 690°
Merck Reference: 14,8513
UN No.: 3084
MDL No.: MFCD00003411
Formel:
AgF₂ Schmelzpunkt: 690 °C Dichte: 4,6 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003411 CAS-Nummer: 7783-95-1 EINECS: 232-037-5 UN-Nummer: 3084 ADR: 8,II Merck Index: 14,8513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...