Gold(I)chlorid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
43365.03
43365.06
43365.03EA
296
EUR
43365.03
43365.06
Gold(I)chlorid ≥99,9% (Metall-Basis)
Gold(I)chlorid
Purity: 99.9%
CAS No.: 10294-29-8
Molecular Formula: AuCl
Formula Weight: 232.42
Physical Form: Powder
Density: 7.57
Melting Point: 289° dec.
Merck Reference: 14,4515
UN No.: 3260
MDL No.: MFCD00046175
Formel:
AuCl Schmelzpunkt: 289 °C (Zersetzung) Dichte: 7,57 |
MDL Number:
MFCD00046175 CAS-Nummer: 10294-29-8 EINECS: 233-655-8 UN-Nummer: 3260 ADR: 8,III Merck Index: 14,4515 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...