Kupfer(II)bromid Cu ≥ 28,1%, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cupric bromide, Cuprous bromide
14258.22
14258.36
14258.22EA
78.7
EUR
14258.22
14258.36
Kupfer(II)bromid Cu ≥ 28,1%, Reagenzqualität
Kupfer(II)bromid
CAS No.: 7789-45-9
Molecular Formula: CuBr2
Formula Weight: 223.36
Physical Form: Crystalline
Density: 4.710
Sensitivity: Hygroscopic
Appearance: Dark grey to black
Odour: Odourless
Melting Point: 498°
Boiling Point: 900°
Merck Reference: 14,2629
UN No.: 3260
MDL No.: MFCD00010970
Formel:
Br₂Cu Siedepunkt: 900 °C Schmelzpunkt: 498 °C Dichte: 4.710 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010970 CAS-Nummer: 7789-45-9 EINECS: 232-167-2 UN-Nummer: 3260 ADR: 8,III Merck Index: 14,2629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...