Aluminiumlanthan(III)oxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Synonyme:
Lanthanum(III) aluminum oxide, Lanthanum Aluminium oxide, Aluminium lanthanum trioxide
39250.06
39250.14
39250.06EA
98.7
EUR
39250.06
39250.14
Aluminiumlanthan(III)oxid ≥99,9% (Metall-Basis)
Aluminiumlanthan(III)oxid
Purity: 99.9%
CAS No.: 12003-65-5
Molecular Formula: LaAlO3
Formula Weight: 213.88
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00144550
Formel:
AlLaO₃ Molekulargewicht: 213.89 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00144550 CAS-Nummer: 12003-65-5 EINECS: 234-433-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...