Ytterbium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
36252.14
36252.22
36252.14EA
239
EUR
36252.14
36252.22
Ytterbium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Ytterbium(III)oxid
Purity: 99.9%
CAS No.: 1314-37-0
Molecular Formula: Yb2O3
Formula Weight: 394.08
Physical Form: 3-12mm Sintered Lump
Density: 9.2
Melting Point: 2227°
Merck Reference: 14,10105
MDL No.: MFCD00011290
Formel:
O₃Yb₂ Schmelzpunkt: 2227 °C Dichte: 9.2 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011290 CAS-Nummer: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...