Neodym(III)oxid ≥99,9%

Lieferant: Thermo Fisher Scientific

Synonyme: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

36275.14 36275.22
36275.14EA 148 EUR
36275.14 36275.22
Neodym(III)oxid ≥99,9%
Neodym(III)oxid

Purity: 99.9% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: 3-12mm Sintered Lump Density: 7.24 Melting Point: 2272° Merck Reference: 14,6450 MDL No.: MFCD00011134

Formel: Nd₂O₃
Siedepunkt: 3760 °C (1013 hPa)
Schmelzpunkt: 2272 °C
Dichte: 7.24
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011134
CAS-Nummer: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR