Europium(III)oxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

203262-25G 203262-5G
SIAL203262-25GEA 1160 EUR
SIAL203262-25G SIAL203262-5G
Europium(III)oxid, Sigma-Aldrich®
Europium(III)oxid
Formel: Eu₂O₃
Schmelzpunkt: 2325 °C
Dichte: 7.42 g/cm³ (20 °C)
MDL Number: MFCD00010997
CAS-Nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR