Dysprosiumtrichlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ quality level = MQ100
325546-25G
325546-5G
SIAL325546-25GEA
982
EUR
SIAL325546-25G
SIAL325546-5G
Dysprosiumtrichlorid, Sigma-Aldrich®
Dysprosiumtrichlorid
Formel:
Cl₃Dy Schmelzpunkt: 642…652 °C Dichte: 3.67 g/cm³ (0 °C) |
MDL Number:
MFCD00010984 CAS-Nummer: 10025-74-8 EINECS: 233-039-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...