5-Nitroindol 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Nitro-1H-indole
128700050
128700250
ACRO128700050EA
76
EUR
ACRO128700050
ACRO128700250
5-Nitroindol 99%
5-Nitroindol
Formel:
C₈H₆N₂O₂ Schmelzpunkt: 140…142 °C |
MDL Number:
MFCD00005673 CAS-Nummer: 6146-52-7 EINECS: 228-153-0 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 140 to 143 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...