5-Indolcarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-Indole-5-carboxylic acid

A18627.03 A18627.06 A18627.14
A18627.03EA 51 EUR
A18627.03 A18627.06 A18627.14
5-Indolcarbonsäure ≥98%
5-Indolcarbonsäure

Purity: 98% CAS No.: 1670-81-1 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 208-212° Beilstein Reference: 124391

Formel: C₉H₇NO₂
Molekulargewicht: 161.16 g/mol
Schmelzpunkt: 208…212 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005678
CAS-Nummer: 1670-81-1
EINECS: 216-799-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR