6-Chlorindol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23650.03
B23650.06
B23650.03EA
21.8
EUR
B23650.03
B23650.06
6-Chlorindol ≥99%
6-Chlorindol
Purity: 99%
CAS No.: 17422-33-2
Molecular Formula: C8H6ClN
Formula Weight: 151.60
Melting Point: 87-90°
Beilstein Reference: 112345
Formel:
C₈H₆ClN Schmelzpunkt: 87…90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005681 CAS-Nummer: 17422-33-2 EINECS: 241-449-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...