Diallyldisulfid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Diallyldisulphide, Allyl disulphide
M-Clarity™ quality level = MQ100
317691-100G
317691-25G
SIAL317691-100GEA
148
EUR
SIAL317691-100G
SIAL317691-25G
Diallyldisulfid, Sigma-Aldrich®
Diallyldisulfid
Formel:
C₆H₁₀S₂ Siedepunkt: 198 °C (1013 hPa) Dichte: 1.0237 g/cm³ (15 °C) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00008656 CAS-Nummer: 2179-57-9 EINECS: 218-548-6 UN-Nummer: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...