Thionylbromid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Thionyl dibromide
L15648.14
L15648.22
L15648.14EA
49.7
EUR
L15648.14
L15648.22
Thionylbromid ≥97%
Thionylbromid
Purity: 97%
CAS No.: 507-16-4
Molecular Formula: Br2OS
Formula Weight: 207.88
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6750
Odour: Acrid
Melting Point: -52°
Boiling Point: 47-49°/20mm
Merck Reference: 14,9347
UN No.: 3264
MDL No.: MFCD00011448
Formel:
Br₂OS Siedepunkt: 47…49 °C (20 mmHg) Schmelzpunkt: –52 °C Dichte: 2.683 Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00011448 CAS-Nummer: 507-16-4 EINECS: 208-064-3 UN-Nummer: 3264 ADR: 8,II Merck Index: 14,9347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...