di-Ammoniumhydrogenphosphat

Lieferant: Roth Carl

Synonyme: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

P736.1 P736.2
ROTHP736.1EA 84 EUR
ROTHP736.1 ROTHP736.2
di-Ammoniumhydrogenphosphat
di-Ammoniumhydrogenphosphat
Formel: (NH₄)₂HPO₄
Molekulargewicht: 132.06 g/mol
Schmelzpunkt: 185
Dichte: 1.619 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010891
CAS-Nummer: 7783-28-0
EINECS: 231-987-8
REACH-Nummer: 01-2119490974-22
Merck Index: 13,00546

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR