Ammoniumiodid ≥99,0% ACS

Lieferant: Thermo Fisher Scientific
Achtung

11668.22 11668.36
11668.22EA 85.2 EUR
11668.22 11668.36
Ammoniumiodid ≥99,0% ACS
Ammoniumiodid

Purity: 99.0% min CAS No.: 12027-06-4 Molecular Formula: NH4I Formula Weight: 144.94 Physical Form: Granular Density: 2.514 Refractive Index: 1.7031 Sensitivity: Light and Air Sensitive and Hygroscopic Appearance: White to pale yellow Odour: Odourless Melting Point: 551° subl. Solubility: Freely soluble in water, alcohol, glycerol, methanol Merck Reference: 14,529 MDL No.: MFCD00011424

Formel: NH₄I
Molekulargewicht: 144.94 g/mol
Schmelzpunkt: 551 °C (Subl.)
Dichte: 2.514
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011424
CAS-Nummer: 12027-06-4
EINECS: 234-717-7
Merck Index: 14,529

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR