Ammoniumchlorid ≥99,5% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammonium muriate, Salmiac

40193.A1 40193.A7
40193.A1EA 75.8 EUR
40193.A1 40193.A7
Ammoniumchlorid ≥99,5% ACS
Ammoniumchlorid

Purity: 99.5% min CAS No.: 12125-02-9 Molecular Formula: NH4Cl Formula Weight: 53.49 Physical Form: Crystalline Density: ≈1.53 Refractive Index: 1.642 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 340° subl. Merck Reference: 14,509 MDL No.: MFCD00011420

Formel: NH₄Cl
Molekulargewicht: 53.49 g/mol
Schmelzpunkt: 340 °C (Subl.)
Dichte: 1.53 g/cm³
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011420
CAS-Nummer: 12125-02-9
EINECS: 235-186-4
Merck Index: 14,509
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR