Acetylthiocholiniodid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
Purity: 98%
CAS No.: 1866-15-5
Molecular Formula: CH3COSCH2CH2N(CH3)3I
Formula Weight: 289.18
Storage Temperature: 2°C to 8°C
Melting Point: 205-209°
Beilstein Reference: 3916578
UN No.: 2811
MDL No.: MFCD00011819
Formel:
H₃CC(S)O(CH₂)₂N(CH₃)₃I Schmelzpunkt: 205…209 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00011819 CAS-Nummer: 1866-15-5 EINECS: 217-474-1 UN-Nummer: 2811 ADR: 6.1,III |