Arsen(III)sulfid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Arsenic trisulphide, Disrsenic trisulphide
43340.14
43340.06
43340.14EA
273
EUR
43340.14
43340.06
Arsen(III)sulfid ≥99,9% (Metall-Basis)
Arsen(III)sulfid
Purity: 99.9%
CAS No.: 1303-33-9
Molecular Formula: As2S3
Formula Weight: 246.03
Physical Form: 1-6mm Fused Pieces
Density: 3.43
Melting Point: 300°
Boiling Point: 707°
Merck Reference: 14,806
UN No.: 1557
MDL No.: MFCD00003435
For vacuum deposition
Formel:
As₂S₃ Siedepunkt: 707 °C Schmelzpunkt: 300 °C Dichte: 3.43 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003435 CAS-Nummer: 1303-33-9 EINECS: 215-117-4 UN-Nummer: 1557 ADR: 6.1,III Merck Index: 14,806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...