Bismut(III)oxid ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bismuth Trioxide, Dibismuth trioxide
42863.18
42863.30
42863.A1
42863.18EA
149
EUR
42863.18
42863.30
42863.A1
Bismut(III)oxid ≥99,999% (Metall-Basis), Puratronic®
Bismut(III)oxid
Purity: 99.999%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: Powder
Density: 8.9
Appearance: Yellow
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462
Formel:
Bi₂O₃ Siedepunkt: ∼1890 °C Schmelzpunkt: 817 °C Dichte: 8.9 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003462 CAS-Nummer: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...