Antimon(III)oxid 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

213470010 213471000 213470051
ACRO213470010EA 173 EUR
ACRO213470010 ACRO213471000 ACRO213470051
Antimon(III)oxid 99+%
Antimon(III)oxid
Formel: O₃Sb₂
Siedepunkt: 1550 °C (760 mmHg)
Schmelzpunkt: 656 °C
Dichte: 5,2 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011214
CAS-Nummer: 1309-64-4
EINECS: 215-175-0
ADR: 6.1,III
Merck Index: 13,00717

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Assay ≥99.0 % (Sb2O3)
Particle size ≤0.01 % Residue on 45 µm sieve
Average particle size 0.8 to 1.0 µm
Arsenic (As) ≤500 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤400 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR