Antimon(III)fluorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Antimony trifluoride
A14068.0B
A14068.18
A14068.30
A14068.0BEA
292
EUR
A14068.0B
A14068.18
A14068.30
Antimon(III)fluorid ≥98%
Antimon(III)fluorid
Purity: 98%
CAS No.: 7783-56-4
Molecular Formula: SbF3
Formula Weight: 178.75
Odour: Odourless
Melting Point: 292°
Boiling Point: 376°
Merck Reference: 14,709
UN No.: 2923
MDL No.: MFCD00011218
Formel:
F₃Sb Siedepunkt: 376 °C Schmelzpunkt: 292 °C Dichte: 4.379 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011218 CAS-Nummer: 7783-56-4 EINECS: 232-009-2 UN-Nummer: 2923 ADR: 8,II Merck Index: 14,709 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...