Antimon(III)bromid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Antimony tribromide

41573.18 41573.30
41573.18EA 173 EUR
41573.18 41573.30
Antimon(III)bromid 99%
Antimon(III)bromid

Purity: 99% CAS No.: 7789-61-9 Molecular Formula: SbBr3 Formula Weight: 361.48 Physical Form: Powder Density: 4.148 Refractive Index: 1.74 Sensitivity: Moisture Sensitive Odour: Characteristic Melting Point: 96.6° Boiling Point: 280° Merck Reference: 14,706 UN No.: 3260 MDL No.: MFCD00016317

Formel: Br₃Sb
Siedepunkt: 280 °C
Schmelzpunkt: 96.6 °C
Dichte: 4.148
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016317
CAS-Nummer: 7789-61-9
EINECS: 232-179-8
UN-Nummer: 3260
ADR: 8,II
Merck Index: 14,706

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR