Zinn(II)sulfid ≥99,5%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tin sulphide

14051.06 14051.14
14051.06EA 161 EUR
14051.06 14051.14
Zinn(II)sulfid ≥99,5%
Zinn(II)sulfid

Purity: 99.5% CAS No.: 1314-95-0 Molecular Formula: SnS Formula Weight: 150.75 Physical Form: -4 Mesh Density: 5.08 Appearance: Grey to black Melting Point: 882° Solubility: Soluble in conc. HCl, hot conc. H2SO4 Merck Reference: 14,8791 MDL No.: MFCD00011245


Polymerization catalyst

Formel: SSn
Schmelzpunkt: 882 °C
Dichte: 5.08
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011245
CAS-Nummer: 1314-95-0
EINECS: 215-248-7
Merck Index: 14,8791

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR