Blei(II)carbonat basisch, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
M-Clarity™ quality level = MQ100
243582-100G
243582-2.5KG
SIAL243582-100GEA
48.3
EUR
SIAL243582-100G
SIAL243582-2.5KG
Blei(II)carbonat basisch, Sigma-Aldrich®
Blei(II)carbonat basisch
Formel:
(PbCO₃)₂·Pb(OH)₂ Schmelzpunkt: 400 °C Dichte: 6.06 g/cm³ (20 °C) |
MDL Number:
MFCD00078155 CAS-Nummer: 1319-46-6 EINECS: 215-290-6 UN-Nummer: 2291 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...