Siliciumcarbid, β-Phase ≥99,8% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

14165.22 14165.36 14165.A3
14165.22EA 110 EUR
14165.22 14165.36 14165.A3
Siliciumcarbid, β-Phase ≥99,8% (Metall-Basis)
Siliciumcarbid, β-Phase

Purity: 99.8% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 1 Micron Powder, S.A. 14-19m2/g Odour: Odourless Merck Reference: 14,8492 MDL No.: MFCD00049531

Formel: SiC
Schmelzpunkt: 2700 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00049531
CAS-Nummer: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR